Measuring scoops
: KARTELL
PP, white. With flat, bevelled end and reinforced handle.
- Excellent chemical resistance, unbreakable
- Heat resistant up to 130 °C, autoclavable
- The capacity of the scoop is stamped into the handle for easy reading
PP, white. With flat, bevelled end and reinforced handle.
- Excellent chemical resistance, unbreakable
- Heat resistant up to 130 °C, autoclavable
- The capacity of the scoop is stamped into the handle for easy reading
Product Family Options
- CapacityLength