Medical scissors, straight
Sharp/sharp, 130 mm
Catalog # 233-0073
Supplier: HAMMACHER KARL
Jump to:
Product Details & Documents
Stainless steel.
- Sharp-sharp
- Corrosion-resistant
- Autoclavable
Specifications
- Type:Sharp/sharp
- Length:130 mm