ContactUsLinkComponent
Minor skin graft blade, No. SG3
Minor skin graft blade, No. SG3
  233-0053
undefined
Minor skin graft blade, No. SG3
  233-0053
  • Description:
    Scalpel blades, special
  • Pk:
    20
Stainless steel. Originally developed for use with skin grafts, these blades are also suitable for a wide range of other applications.
  • Fits standard No. 3 surgical handle (233-5454)
  • Sterile
  • Individually packed
Ideal for use on the hands and feet of small children given its practical size advantage compared to a standard skin grafting knife. Also suitable for use within Histology departments for the fine sectioning of the pathology blocks.
 : Packed in individual foil packs.
Handle