O-rings for flat flanges, DURAN®
Supplier: DWK Life Sciences
Certificates
About this item
Silicone core sheathed with FEP, red.
- Guarantee excellent joint seal
- Temperature resistant from −200 to +200 °C
- Excellent chemical resistance
O-rings for flat flanges with groove.