VWR®, Test Tube Racks
Supplier: VWR Collection
Certificates
About this item
PTFE.
- Excellent chemical resistance
- Temperature resistant from –200 to +280 °C
- Non stick surfaces for easy cleaning
Robust and inert to virtually all chemical reagents, with very good thermal resistance. Suitable for use in cryogenics, freezers and ovens.