Pasteur pipettes, VOLAC®
Supplier: Poulten & Graf
Jump to:
About this item
Glass, graduated, plain or pre-plugged, sterile or non sterile.
- Precision pipettes with a heavy wall thickness to provide the best quality product allied with greater strength to give lower breakages during transit, and whilst in use or, when fitting PVC teats
- Standard pipettes with top body, outer diameter 6,8 to 7,1 mm, wall thickness 0,50 to 0,53 mm and jet outer diameter 1,50 ±0,2 mm
4 Options Available Below
Product Details & Documents
Glass, graduated, plain or pre-plugged, sterile or non sterile.
- Precision pipettes with a heavy wall thickness to provide the best quality product allied with greater strength to give lower breakages during transit, and whilst in use or, when fitting PVC teats
- Standard pipettes with top body, outer diameter 6,8 to 7,1 mm, wall thickness 0,50 to 0,53 mm and jet outer diameter 1,50 ±0,2 mm
Product Family Options
Standard, unplugged and non sterile
Product Information
Standard, pre-plugged and non sterile
Product Information
Accessories
Product Information
- Catalog #AvailabilityPrice
- Pipette teat, PVC, 1,5 ml