VWR®, PCR Plates, 96 Well
Supplier: VWR Collection
Certificates
About this item
PP. These PCR plates are compatible with most thermal cyclers.
- Smooth, thin, uniform well walls ensure accurate thermal transfer
- Plates are thin and flexible
- Certified free from DNase, RNase and human genomic DNA
- Printed alphanumeric labelling and cut corner simplifies plate orientation and sample identification
Working capacity: 200 µl