PCR plates, 24- and 48-wells, Thermo-Fast®
PP. Suitable for 0,2 ml thermal cycler blocks.
- Sealable using adhesive films and foils, heat seals and flat or domed cap strips
- Maximum well volume 0,3 ml when used with adhesive and heat seals
- Certified free from DNase, RNase and human genomic DNA
Colour: Natural