PCR plates, 96-well, semi-skirted, Thermo-Fast®
Supplier: Thermo Fisher Scientific
Certificates
About this item
PP. Compatible with all major thermal cyclers.
- Greater rigidity than non-skirted plates and provides space for barcoding
- Segmented design allows it to be cut into 24 and 48 well sections
- Maximum well volume 0,3 ml when used with adhesive and heat seals
- Cut corner (A12)