Orders LinkContactUsLinkComponent
PCR plates
Supplier:   Simport Scientific
PCR plates
Supplier:  Simport Scientific
Certificates
PCR plates
15 Options Available Below
Product Details & Documents
PP. Thin walled PCR plates compatible with 0,2 ml thermal cyclers. The insides of the tubes are smooth and have an inert surface on which enzymes and nucleic acids do not bind.
  • Alphanumeric grid for better identification
  • Flexible plate for better fitting of tubes in the thermal block
  • Larger plates are easily cut into sections of 24, 32 or 48 tubes, but, for more convenience, pre-cut plates are also available with formats of 48 tubes (6×8) and 24 tubes (3×8)
  • The bottom right corner of the plate is cut away to facilitate orientation
  • Certified free of RNase, DNase and DNA
 : Packed in tamperproof, resealable bags of 10 plates.
Product Family Options
Product Information
  • No. of wells
    Skirt
    Well colour
    Availability
    Price
  • 24
    Non-skirted
    Clear
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 85,90 €
    85,90 €
    1 * 50 ST
       85,90 €
  • 24
    Non-skirted
    Blue
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 85,90 €
    85,90 €
    1 * 50 ST
       85,90 €
  • 24
    Non-skirted
    Green
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 85,90 €
    85,90 €
    1 * 50 ST
       85,90 €
  • 24
    Non-skirted
    Red
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 85,90 €
    85,90 €
    1 * 50 ST
       85,90 €
  • 24
    Non-skirted
    Yellow
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 85,90 €
    85,90 €
    1 * 50 ST
       85,90 €
  • 48
    Non-skirted
    Blue
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 127,00 €
    127,00 €
    1 * 50 ST
       127,00 €
  • 48
    Non-skirted
    Green
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 127,00 €
    127,00 €
    1 * 50 ST
       127,00 €
  • 48
    Non-skirted
    Clear
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 127,00 €
    127,00 €
    1 * 50 ST
       127,00 €
  • 48
    Non-skirted
    Red
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 127,00 €
    127,00 €
    1 * 50 ST
       127,00 €
  • 48
    Non-skirted
    Yellow
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 127,00 €
    127,00 €
    1 * 50 ST
       127,00 €
  • 96
    Non-skirted
    Blue
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 539,00 €
    539,00 €
    1 * 100 ST
       539,00 €
  • 96
    Non-skirted
    Green
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 539,00 €
    539,00 €
    1 * 100 ST
       539,00 €
  • 96
    Non-skirted
    Clear
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 539,00 €
    539,00 €
    1 * 100 ST
       539,00 €
  • 96
    Non-skirted
    Red
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 539,00 €
    539,00 €
    1 * 100 ST
       539,00 €
  • 96
    Non-skirted
    Yellow
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 539,00 €
    539,00 €
    1 * 100 ST
       539,00 €