PCR plates, low profile
211-3382
- No. of wells:96
- Profile:Low
- Skirt:Non-skirted
- Well colour:Clear
- Pk:100
PP. Thin walled 96-well PCR plates with small tube volume (100 µl).
- Alphanumeric grid for easy sample identification
- Flat surface for better sealing
- Can be cut to desired format
- Small volume reducing dead space between sample and cover
: Packed in tamperproof, resealable bags of 10 plates.