PCR plates, skirted
PP. These thin-walled PCR plates are suitable for robotic handling, and are ideal for use with automated pipetting systems.
- Each well of the 96-well plate can hold a volume of 100 µl, while the 384-well plate has a capacity of 40 µl per well, suitable for reaction volumes of between 2 and 30 µl
- Area for bar coding or labelling on each side and top
- Alphanumeric grid for easy sample identification
- Flat surface for better sealing
: Packed in tamperproof, resealable bags of 10 plates.