ContactUsLinkComponent
PCR plates, 384-well, Thermo-Fast®
PCR plates, 384-well, Thermo-Fast®
Catalog # 732-4961
undefined
PCR plates, 384-well, Thermo-Fast®
Catalog # 732-4961
Specifications
  • No. of wells:
    384
  • Skirt:
    Fully skirted
  • Well colour:
    Clear
  • Pk:
    50
Specifications
About this item
PP. Designed for high throughput PCR, this 384-well plate is compatible with the majority of 384 block PCR, qPCR and sequencing instruments.
  • The full skirt and rigid plate deck minimise distortion during PCR making it suitable for use with robotic systems
  • Eight holes in the skirt aid plate positioning and removal from the thermal cycler block
  • Certified free from DNase, RNase and human genomic DNA
  • Alphanumeric grid referencing and cut away corners aid orientation
Maximum well volume: 40 µl
Working volume: 25 µl