Storage plate, 48-well, 6,0 ml
732-4923
- Colour:Natural
- Description:Storage plate, 48-well, 6,0 ml
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Pk:50
PP. 48 square well plates for sample storage.
- 6,0 ml working volume when sealed with adhesive or heat seals
- Pyramidal well bottom for ease of sample retrieval
- Support and stacking ribs
- Sealing options include heat sealing, gas permeable adhesive seals, adhesive sealing, and sealing mats
Colour: Natural