Cell culture insert plates, ThinCert™
About this item
PS, sterile, with lid. Designed for use in air-lift culture where tissue is cultivated in permeable membrane supports (cell culture inserts), with the cell culture medium nourishing the cells from below the membrane and the air exerting its influence from above. If cell culture inserts and conventional cell culture plates are utilised in the air-lift culture, the available volume of culture medium is severely limited. This reduced volume results in an elevated nutrient consumption rate from the lower compartment and an increased number of medium changes.
- Optimised for use with ThinCert™ cell culture inserts
- Deep wells for an increased volume of medium in air-lift culture
- Notches for fixed insert position
- Available in 6 and 12 well format
With the ThinCert™ plate the deep wells allow a larger volume of medium to be available to the culture. As a consequence, fewer medium changes are required and the amount of manual work is reduced. The reduced frequency of medium changes ensures that the medium conditioning lasts longer and an improved tissue quality is achieved.
2 Options Available Below
Product Details & Documents
PS, sterile, with lid. Designed for use in air-lift culture where tissue is cultivated in permeable membrane supports (cell culture inserts), with the cell culture medium nourishing the cells from below the membrane and the air exerting its influence from above. If cell culture inserts and conventional cell culture plates are utilised in the air-lift culture, the available volume of culture medium is severely limited. This reduced volume results in an elevated nutrient consumption rate from the lower compartment and an increased number of medium changes.
- Optimised for use with ThinCert™ cell culture inserts
- Deep wells for an increased volume of medium in air-lift culture
- Notches for fixed insert position
- Available in 6 and 12 well format
With the ThinCert™ plate the deep wells allow a larger volume of medium to be available to the culture. As a consequence, fewer medium changes are required and the amount of manual work is reduced. The reduced frequency of medium changes ensures that the medium conditioning lasts longer and an improved tissue quality is achieved.
Specifications for Product Family
Specifications
- Catalog No:
- 392-0035
- 392-0048
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
Specifications
Product Family Options
Product Information
- VolumeAvailabilityPrice
- 392-00356-well plate, 30 ml (20 ml working volume in air-lift culture)6-well plate, 30 ml (20 ml working volume in air-lift culture)Log in for availability1 * 50 ST1 050,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.392-0035Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact - 392-004812-well plate, 10 ml (4 ml working volume in air-lift culture)12-well plate, 10 ml (4 ml working volume in air-lift culture)10 ml (4 ml working volume in air-lift culture)Log in for availability1 * 60 ST1 110,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.392-0048Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (241002); ISO 50001 (241002)Low Manufacturing Impact