ContactUsLinkComponent
Sterilin® Microtitre Plates, Black, 96-Well
Sterilin® Microtitre Plates, Black, 96-Well
  634-0006
 :  Sterilin
undefined
Sterilin® Microtitre Plates, Black, 96-Well
  634-0006
 :  Sterilin
  • Form:
    8×12
  • No. of wells:
    96
  • Sterile:
    -
  • Colour:
    Black
  • Description:
    96-well plate, flat-bottomed, black
  • Pk:
    50
  • Well volume:
    400 µl
PS, non-treated. Ideal for fluorescence work and bioluminescence studies.
  • Black 96-well plates are ideal for fluorescence work where the pigment prevents cross-talk between the wells - low background fluorescence minimises light scattering
  • SBS compliant, ensuring suitable fit with all automation
  • Manufactured from virgin polystyrene
  • Supplied nonsterile, providing a medium binding surface
  • Chimney well design to help reduce contamination
  • Supplied with no lid
Colour: Black