Order Entry
Germany
ContactUsLinkComponent
Gas tubing for gas burners
Gas tubing for gas burners
Catalog # 451-4330
Supplier:  DEUTSCH NEUMANN
undefined
Gas tubing for gas burners
Catalog # 451-4330
Supplier:  DEUTSCH NEUMANN

Specifications

  • Tubing ext.Ø:
    14 mm
  • Tubing int.Ø:
    10 mm
  • Wall thickness:
    2 mm
  • Pk:
    1 m

Specifications

About this item

Red rubber gas tubing for gas burners, without sheath or armouring, in line with DIN 30 664. For all gases in line with DVGW process sheet G 260.

Operating pressure up to 100 mbar