ContactUsLinkComponent
General purpose gloves, Resistex Light Grip 2
General purpose gloves, Resistex Light Grip 2
Supplier:  HONEYWELL SAFETY
Certificates
undefined
General purpose gloves, Resistex Light Grip 2
Supplier:  HONEYWELL SAFETY
Specifications
  • Catalog No:
  • 112-4049
  • 112-4051
  • 112-4050
  • 112-4052
  • Model:
  • Resistex Light Grip 2
  • Resistex Light Grip 2
  • Resistex Light Grip 2
  • Resistex Light Grip 2
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Polyester/cotton
  • Polyester/cotton
  • Polyester/cotton
  • Polyester/cotton
  • Finish:
  • Dot coating on both sides
  • Dot coating on both sides
  • Dot coating on both sides
  • Dot coating on both sides
  • Gauge:
  • 13
  • 13
  • 13
  • 13
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (113XX)
  • EN 388:2016 (113XX)
  • EN 388:2016 (113XX)
  • EN 388:2016 (113XX)
Specifications