Chemical resistant gloves, AlphaTec® 87-950 / 87-955
Supplier: Ansell
Certificates
About this item
These cotton flock lined and chlorinated latex gloves with high perspiration absorbance are comfortable to wear and feature a embossed lozenge finish for a better grip.
- Excellent resistance to mechanical hazards and increased chemical protection
- Extremely high resistance to many ketones, salts, detergents, alcohols, alkalis and fats
- Chlorinated: For better grip and increased chemical resistance
Suitable for heavy duty handling, chemical and petrol processing and maintenance activities.
Certifications: EN ISO 374-1 Type A (AKLOPS), EN ISO 374-5 VIRUS, EN 388:2016 (X121X); Cat. III
Specifications
- Catalog No:
- 112-0311
- 112-0309
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Finish:
- Lozenge
- Lozenge
- Cuff style:
- Pinked
- Pinked
- EN norm:
- EN ISO 374-1 Type A (AKLOPS), EN ISO 374-5 VIRUS, EN 388:2016 (X121X)
- EN ISO 374-1 Type A (AKLOPS), EN ISO 374-5 VIRUS, EN 388:2016 (X121X)