Cleanroom gloves, nitrile, AdvanTech 529
About this item
These ambidextrous, liquidproof gloves with beaded cuffs are made of nitrile.
- Optimal thickness providing superior dexterity and ease of movement
- 100% nitrile formulation eliminating protein allergy concerns
- Lightly textured fingertips for an enhanced wet and dry grip
- Long length to keep glove up on garment sleeve
The nitrile formulation provides superior mechanical resistance and splash protection against chemicals used in controlled environments. Free of pigments, powder, silicone or paraffin. Ideal for use in pharmaceutical industries, medical device manufacturing or microchip assembly.
Suitable for use in Class 100, ISO 5 or M3.5 cleanrooms.
Certifications: EN ISO 374-1 Type B (JKT), EN ISO 374-5, EN 421, Cat. III; AQL 1,5.
Packaging: Packaged in Class 10/M2.5 compatible, IPA resistant cleanroom bags. Lot traceability on inner bags and cartons.
5 Options Available Below
Product Details & Documents
These ambidextrous, liquidproof gloves with beaded cuffs are made of nitrile.
- Optimal thickness providing superior dexterity and ease of movement
- 100% nitrile formulation eliminating protein allergy concerns
- Lightly textured fingertips for an enhanced wet and dry grip
- Long length to keep glove up on garment sleeve
The nitrile formulation provides superior mechanical resistance and splash protection against chemicals used in controlled environments. Free of pigments, powder, silicone or paraffin. Ideal for use in pharmaceutical industries, medical device manufacturing or microchip assembly.
Suitable for use in Class 100, ISO 5 or M3.5 cleanrooms.
Certifications: EN ISO 374-1 Type B (JKT), EN ISO 374-5, EN 421, Cat. III; AQL 1,5.
Packaging: Packaged in Class 10/M2.5 compatible, IPA resistant cleanroom bags. Lot traceability on inner bags and cartons.
Specifications for Product Family
Specifications
- Catalog No:
- 112-0726
- MAPA34529007
- 112-0728
- 112-0729
- 112-0725
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cleanroom class:
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- EN norm:
- EN ISO 374-1 Type B (JKT), EN ISO 374-5, EN 421
- EN ISO 374-1 Type B (JKT), EN ISO 374-5, EN 421
- EN ISO 374-1 Type B (JKT), EN ISO 374-5, EN 421
- EN ISO 374-1 Type B (JKT), EN ISO 374-5, EN 421
- EN ISO 374-1 Type B (JKT), EN ISO 374-5, EN 421
- Anti-static:
- +
- +
- +
- +
- +
Specifications
Product Family Options
Product Information
- Glove sizeColourLengthThicknessPackedAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.112-0726Ambidextrous / hand-specificAmbidextrousDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowder-freeCleanroom class100/ISO 5Cuff styleBeadedEN normEN ISO 374-1 Type B (JKT), EN ISO 374-5, EN 421Anti-static+ - Specifications:
More
Specifications
Cat. No.MAPA34529007Ambidextrous / hand-specificAmbidextrousDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowder-freeCleanroom class100/ISO 5Cuff styleBeadedEN normEN ISO 374-1 Type B (JKT), EN ISO 374-5, EN 421Anti-static+ - Specifications:
More
Specifications
Cat. No.112-0728Ambidextrous / hand-specificAmbidextrousDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowder-freeCleanroom class100/ISO 5Cuff styleBeadedEN normEN ISO 374-1 Type B (JKT), EN ISO 374-5, EN 421Anti-static+ - Specifications:
More
Specifications
Cat. No.112-0729Ambidextrous / hand-specificAmbidextrousDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowder-freeCleanroom class100/ISO 5Cuff styleBeadedEN normEN ISO 374-1 Type B (JKT), EN ISO 374-5, EN 421Anti-static+ - Specifications:
More
Specifications
Cat. No.112-0725Ambidextrous / hand-specificAmbidextrousDisposable / reusableDisposableLatex / latex-freeLatex-freePowdered / powder-freePowder-freeCleanroom class100/ISO 5Cuff styleBeadedEN normEN ISO 374-1 Type B (JKT), EN ISO 374-5, EN 421Anti-static+