Dropping funnels, cylindrical, with pressure balance
DURAN®, borosilicate glass, clear, with glass key.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Standard ground joint cone and socket, with divisions for approximate volume reading. Stopcock with thread guard.