Funnel, disposable
Disposable funnel, 60 mm
Catalog # 221-0083
Supplier: MARYLAND PLASTICS
Jump to:
Product Details & Documents
Transparent PP, single use
- Very good chemical resistance
- Autoclavable
- Suitable for use with standard filter paper
Specifications
- Funnel O-Ø:60 mm
- Description:Disposable funnel