Orders LinkContactUsLinkComponent
Forceps for cover glasses
Forceps for cover glasses
Supplier:   BOCHEM
Forceps for cover glasses
Supplier:  BOCHEM
Certificates
Forceps for cover glasses
Forceps for cover glasses
3 Options Available Below
Product Details & Documents
Stainless steel or stainless steel with PTFE coating.
  • For easy, secure handling of cover glasses
  • With 6 mm wide tips
  • Can also be used for other laboratory equipment, such as filter discs
  • PTFE coated design with particularly smooth surface to prevent contamination and enable simple cleaning
Product Family Options
Product Information
  • Length
    Availability
    Price
  • 232-1262
    Cover glass forceps, bent, stainless steel
    Cover glass forceps, bent, stainless steel
    105 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5,70 €
    5,70 €
    1 * 1 ST
       5,70 €
  • 232-0021
    Cover glass forceps, bent, PTFE coating
    Cover glass forceps, bent, PTFE coating
    105 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 29,20 €
    29,20 €
    1 * 1 ST
       29,20 €
  • 232-0060
    Cover glass forceps, straight, stainless steel
    Cover glass forceps, straight, stainless steel
    105 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6,70 €
    6,70 €
    1 * 1 ST
       6,70 €