Vacuum tweezers, aspirette®
Powered by rechargeable battery for picking up and placing flat objects up to a max. unit weight of 70 g.
- One-button operation, ergonomic shape
- Standard and antistatic models available
- Rechargeable battery can be charged ∼1000 times
: The package includes a mains charger and a suction unit set consisting of three suction nozzles with 3,2, 6 and 10 mm Ø suction cups made of silicone.