VWR®, Flasks, Round Bottom, Narrow or Wide Neck
Supplier: VWR Collection
Jump to:
About this item
Borosilicate glass 3,3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
The economical alternative for a wide range of applications. Marked with capacity and a white labelling area.
ISO 1773 (narrow mouth)
DIN 12347 (wide mouth)
: ISO 1773 (narrow neck)
DIN 12347 (wide neck)
10 Options Available Below
Product Details & Documents
Borosilicate glass 3,3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
The economical alternative for a wide range of applications. Marked with capacity and a white labelling area.
ISO 1773 (narrow mouth)
DIN 12347 (wide mouth)
: ISO 1773 (narrow neck)
DIN 12347 (wide neck)
Product Family Options
Narrow neck
Product Information
- CapacityHeightExt.ØNeck Ø ext.AvailabilityPrice
Wide neck
Product Information
- CapacityHeightExt.ØNeck Ø ext.AvailabilityPrice
Cork rings for round bottom flasks
Product Information
- Catalog #Ext.ØForAvailabilityPrice