Orders LinkContactUsLinkComponent
Rotary evaporator receiving flasks
Rotary evaporator receiving flasks
Supplier:   Lenz Laborglas
Rotary evaporator receiving flasks
Supplier:  Lenz Laborglas
Certificates
Rotary evaporator receiving flasks
Rotary evaporator receiving flasks
12 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass, clear or plastic-coated.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Spherical joint KS 35/20, centred, suitable for all rotary evaporators.
Dokumente
Product Family Options
Uncoated
Product Information
  • Capacity
    Socket
    Availability
    Price
  • 100 ml
    KS 35/20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 48,20 €
    48,20 €
    1 * 1 ST
       48,20 €
  • 250 ml
    KS 35/20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 50,50 €
    50,50 €
    1 * 1 ST
       50,50 €
  • 500 ml
    KS 35/20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 54,60 €
    54,60 €
    1 * 1 ST
       54,60 €
  • 1000 ml
    KS 35/20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 66,20 €
    66,20 €
    1 * 1 ST
       66,20 €
  • 2000 ml
    KS 35/20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 94,80 €
    94,80 €
    1 * 1 ST
       94,80 €
Coated
Product Information
  • Capacity
    Socket
    Availability
    Price
  • 100 ml
    KS 35/20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 102,00 €
    102,00 €
    1 * 1 ST
       102,00 €
  • 250 ml
    KS 35/20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 105,00 €
    105,00 €
    1 * 1 ST
       105,00 €
  • 500 ml
    KS 35/20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 117,00 €
    117,00 €
    1 * 1 ST
       117,00 €
  • 1000 ml
    KS 35/20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 128,00 €
    128,00 €
    1 * 1 ST
       128,00 €
  • 2000 ml
    KS 35/20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 170,00 €
    170,00 €
    1 * 1 ST
       170,00 €
  • 3000 ml
    KS 35/20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 236,00 €
    236,00 €
    1 * 1 ST
       236,00 €
  • 4000 ml
    KS 35/20
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 249,00 €
    249,00 €
    1 * 1 ST
       249,00 €