Orders LinkContactUsLinkComponent
Eyewash station, HypaClens
Eyewash station, HypaClens
Eyewash station, HypaClens
Eyewash station, HypaClens
This eye care station with mirror includes two HypaClens eyewash bottles (500 ml) filled with 0,9% sterile saline solution. Ideal for irrigating eyes and general wound washing applications.
  • Robust, wall mountable eyewash station
  • Integrated mirror is ideal for self-administration of eyewash
  • Clear instruction panel indicates correct use of eye wash
  • Highly visible design
By twisting off the top the sterile contents may be applied drop by drop or in a continuous gentle stream by squeezing the bottle, a stronger jet may be delivered by breaking off the bottle neck. Rapid pouring of the sterile contents for immediate irrigation or emergency use. The specially designed pouring lip ensures that contents do not touch the sides of the bottle in use.
 : Spare solution bottles can be ordered separately. Wall fixings are not supplied.
Product Family Options
Spare parts
  • Description
  • Eye wash solution, refill bottle, 500 ml
    1 * 1 ST
    1 * 1 ST - 12,00 €
    12,00 €
    1 * 1 ST
       12,00 €
  • Description
  • Eyewash station, complete with two sterile saline solution bottles, mirror and instructions
    1 * 1 KIT
    1 * 1 KIT - 84,60 €
    84,60 €
    1 * 1 KIT
       84,60 €