Personal eyewash
Über diesen Artikel
Sterile, buffered, isotonic sodium chloride solution for rinsing eyes following an accident. Available as a single-use, pocket version (235 ml) or wall mounted (500 ml). The bottle opens automatically when twisted out of the wall bracket, and must therefore, be used immediately.
- Quick and easy to open
- Eye cup protected from dust
- Generous flow of fluid for effective rinsing - the eye cup on the bottle channels the fluid to the eye, avoiding unnecessary spills
- Neutralises the pH value
Expiry date: 4 years if container not opened.
* Bottle holder is supplied without eyewash.
5 mögliche Produktvariationen vorhanden
Product Details & Documents
Sterile, buffered, isotonic sodium chloride solution for rinsing eyes following an accident. Available as a single-use, pocket version (235 ml) or wall mounted (500 ml). The bottle opens automatically when twisted out of the wall bracket, and must therefore, be used immediately.
- Quick and easy to open
- Eye cup protected from dust
- Generous flow of fluid for effective rinsing - the eye cup on the bottle channels the fluid to the eye, avoiding unnecessary spills
- Neutralises the pH value
Expiry date: 4 years if container not opened.
* Bottle holder is supplied without eyewash.
Documents
Product Family Options
Produktinformationen
- AvailabilityPrice
- 111-0164Eyewash solution, sterile, pocket model, 235 ml
- 118-0153Eyewash solution, sterile, 500 ml, twin pack
- 118-0001Wall bracket for 500 ml bottle*
- 118-0248Strap for wall bracket, for extra support for the bottle when located in vibrating environmentsStrap for wall bracket, for extra support for the bottle when located in vibrating environmentsAnmelden für die Verfügbarkeit1 * 1 ST12,70 €
- 111-0165Holster for pocket model, green, with belt loop, 8×4,5×16 mmHolster for pocket model, green, with belt loop, 8×4,5×16 mmAnmelden für die Verfügbarkeit1 * 1 ST12,90 €
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.