Order Entry
Germany
Orders LinkContactUsLinkComponent
Blotting transfer papers

Blotting transfer papers

Supplier:   Ahlstrom
Blotting transfer papers
Supplier:  Ahlstrom
Certificates
Blotting transfer papers

42 Options Available Below

Product Details & Documents

Ahlstrom-Munksjö offers a full range of high quality cotton papers with high absorbency – ideal for chromatographic analysis and blotting methods with gel (Southern, Northern and Western Blots). Ahlstrom-Munksjö manufactures, both in Europe and in the US, absorbent papers made of the highest purity cotton linters. These media do not contain additive of any kind, thus preventing any contamination during the transfer steps.

  • High purity
  • Uniform absorption
  • High intra and inter-lot consistency
  • Wide range of thickness and absorption levels
  • Customised sizing and packaging

Ahlstrom-Munksjö’s absorbent papers are produced with a wide range of thickness and absorption levels for optimal wicking capabilities, drying and blotting in any application. Each media is specifically designed to ensure high quality and consistency. Close surveillance in tightly controlled conditions ensures the highest uniformity, consistency and smoothness of the sheets in all grades. This contributes to clean and even transfers during blotting.

Applications: Chromatography, electrophoresis, Northern, Western and Southern blots, antibiotic discs, seed germination testing.

All references are available in customised sizing and packaging to meet customer needs and industry standards.

Specifications for Product Family

Specifications

  • Catalog No:
  • BINZ2.519.075080-N
  • BINZ2.519.075115-N
  • BINZ143246
  • BINZ160228
  • BINZ143250
  • BINZ143252
  • BINZ143255
  • BINZ143036
  • BINZ143037
  • 512-6134
  • BINZ143038
  • 512-6133
  • BINZ143041
  • 512-6132
  • BINZ143043
  • 512-6139
  • BINZ143046
  • 512-6137
  • BINZ143020
  • 512-6138
  • 512-6136
  • 512-6135
  • 512-6145
  • 514-1001
  • 512-6144
  • BINZ143057
  • 514-1002
  • 514-1003
  • BINZ143025
  • 514-1000
  • 512-6143
  • 514-1006
  • 514-1005
  • BINZ143012
  • 512-6142
  • 512-6140
  • BINZ143047
  • BINZ143269
  • BINZ143271
  • BINZ143273
  • BINZ143287
  • BINZ143296
  • Grade:
  • BF 2
  • BF 2
  • BF 2
  • BF 2
  • BF 2
  • BF 2
  • BF 2
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • 1600
  • BF 3
  • BF 3
  • BF 3
  • BF 4
  • BF 4
  • Weight:
  • 195 g/m²
  • 195 g/m²
  • 195 g/m²
  • 195 g/m²
  • 195 g/m²
  • 195 g/m²
  • 195 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 250 g/m²
  • 330 g/m²
  • 330 g/m²
  • 330 g/m²
  • 550 g/m²
  • 550 g/m²
  • Thickness:
  • 0,35 mm
  • 0,35 mm
  • 0,35 mm
  • 0,35 mm
  • 0,35 mm
  • 0,35 mm
  • 0,35 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,48 mm
  • 0,75 mm
  • 0,75 mm
  • 0,75 mm
  • 1,39 mm
  • 1,39 mm
  • Capillary rise:
  • 65 mm/10 min; 100 mm/30 min
  • 65 mm/10 min; 100 mm/30 min
  • 65 mm/10 min; 100 mm/30 min
  • 65 mm/10 min; 100 mm/30 min
  • 65 mm/10 min; 100 mm/30 min
  • 65 mm/10 min; 100 mm/30 min
  • 65 mm/10 min; 100 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 70 mm/10 min; 110 mm/30 min
  • 130 mm/10 min; 200 mm/30 min
  • 130 mm/10 min; 200 mm/30 min
  • 130 mm/10 min; 200 mm/30 min
  • 160 mm/10 min; 220 mm/30 min
  • 160 mm/10 min; 220 mm/30 min

Specifications

Product Family Options

Grade BF 2

Product Information

  • Dimensions
    Availability
    Price
  • Sheets
    75×80 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 51,40 €
    51,40 €
    1 * 100 ST
       51,40 €

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ2.519.075080-N
    Grade
    BF 2
    Weight
    195 g/m²
    Thickness
    0,35 mm
    Capillary rise
    65 mm/10 min; 100 mm/30 min
  • Sheets
    75×115 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 90,80 €
    90,80 €
    1 * 100 ST
       90,80 €

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ2.519.075115-N
    Grade
    BF 2
    Weight
    195 g/m²
    Thickness
    0,35 mm
    Capillary rise
    65 mm/10 min; 100 mm/30 min
  • Sheets
    200×200 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 88,70 €
    88,70 €
    1 * 100 ST
       88,70 €

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143246
    Grade
    BF 2
    Weight
    195 g/m²
    Thickness
    0,35 mm
    Capillary rise
    65 mm/10 min; 100 mm/30 min
  • Sheets
    205×205 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ160228
    Grade
    BF 2
    Weight
    195 g/m²
    Thickness
    0,35 mm
    Capillary rise
    65 mm/10 min; 100 mm/30 min
  • Sheets
    300×600 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 204,00 €
    204,00 €
    1 * 100 ST
       204,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143250
    Grade
    BF 2
    Weight
    195 g/m²
    Thickness
    0,35 mm
    Capillary rise
    65 mm/10 min; 100 mm/30 min
  • Sheets
    460×570 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 361,00 €
    361,00 €
    1 * 100 ST
       361,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143252
    Grade
    BF 2
    Weight
    195 g/m²
    Thickness
    0,35 mm
    Capillary rise
    65 mm/10 min; 100 mm/30 min
  • Sheets
    580×600 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 409,00 €
    409,00 €
    1 * 100 ST
       409,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143255
    Grade
    BF 2
    Weight
    195 g/m²
    Thickness
    0,35 mm
    Capillary rise
    65 mm/10 min; 100 mm/30 min

Grade 1600

Product Information

  • Dimensions
    Availability
    Price
  • Discs
    Ø 120 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 79,00 €
    79,00 €
    1 * 200 ST
       79,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143036
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Discs
    Ø 125 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 83,20 €
    83,20 €
    1 * 200 ST
       83,20 €

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143037
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Discs
    Ø 140 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 90,00 €
    90,00 €
    1 * 200 ST
       90,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    512-6134
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Discs
    Ø 150 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 97,00 €
    97,00 €
    1 * 200 ST
       97,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143038
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Discs
    Ø 170 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 113,00 €
    113,00 €
    1 * 200 ST
       113,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    512-6133
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Discs
    Ø 180 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 120,00 €
    120,00 €
    1 * 200 ST
       120,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143041
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Discs
    Ø 220 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 145,00 €
    145,00 €
    1 * 200 ST
       145,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    512-6132
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Discs
    Ø 230 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 150,00 €
    150,00 €
    1 * 200 ST
       150,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143043
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Sheets
    120×250 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 94,50 €
    94,50 €
    1 * 200 ST
       94,50 €

    Specifications:
    More
    Specifications
    Cat. No.
    512-6139
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Sheets
    125×125 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 56,50 €
    56,50 €
    1 * 200 ST
       56,50 €

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143046
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Sheets
    140×230 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 122,00 €
    122,00 €
    1 * 200 ST
       122,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    512-6137
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Sheets
    140×289 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 154,00 €
    154,00 €
    1 * 200 ST
       154,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143020
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Sheets
    145×230 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 90,80 €
    90,80 €
    1 * 200 ST
       90,80 €

    Specifications:
    More
    Specifications
    Cat. No.
    512-6138
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Sheets
    150×150 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 59,20 €
    59,20 €
    1 * 200 ST
       59,20 €

    Specifications:
    More
    Specifications
    Cat. No.
    512-6136
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Sheets
    150×350 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 165,00 €
    165,00 €
    1 * 200 ST
       165,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    512-6135
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Sheets
    165×165 mm
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 391,00 €
    391,00 €
    1 * 2.000 ST
       391,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    512-6145
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Sheets
    165×165 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 68,90 €
    68,90 €
    1 * 200 ST
       68,90 €

    Specifications:
    More
    Specifications
    Cat. No.
    514-1001
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Sheets
    170×170 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 70,40 €
    70,40 €
    1 * 200 ST
       70,40 €

    Specifications:
    More
    Specifications
    Cat. No.
    512-6144
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Sheets
    175×175 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 78,80 €
    78,80 €
    1 * 200 ST
       78,80 €

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143057
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Sheets
    200×200 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 105,00 €
    105,00 €
    1 * 200 ST
       105,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    514-1002
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Sheets
    210×297 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 163,00 €
    163,00 €
    1 * 200 ST
       163,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    514-1003
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Sheets
    215×920 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 622,00 €
    622,00 €
    1 * 200 ST
       622,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143025
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Sheets
    219×960 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 737,00 €
    737,00 €
    1 * 200 ST
       737,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    514-1000
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Sheets
    220×945 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 728,00 €
    728,00 €
    1 * 200 ST
       728,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    512-6143
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Sheets
    240×240 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 235,00 €
    235,00 €
    1 * 200 ST
       235,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    514-1006
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Sheets
    290×340 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 322,00 €
    322,00 €
    1 * 200 ST
       322,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    514-1005
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Sheets
    38×80 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 131,00 €
    131,00 €
    1 * 1.000 ST
       131,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143012
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Sheets
    320×320 mm
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 322,00 €
    322,00 €
    1 * 200 ST
       322,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    512-6142
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Sheets
    480×480 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 171,00 €
    171,00 €
    1 * 50 ST
       171,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    512-6140
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min
  • Sheets
    580×600 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 429,00 €
    429,00 €
    1 * 100 ST
       429,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143047
    Grade
    1600
    Weight
    250 g/m²
    Thickness
    0,48 mm
    Capillary rise
    70 mm/10 min; 110 mm/30 min

Grade BF 3

Product Information

  • Dimensions
    Availability
    Price
  • Sheets
    300×600 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 230,00 €
    230,00 €
    1 * 50 ST
       230,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143269
    Grade
    BF 3
    Weight
    330 g/m²
    Thickness
    0,75 mm
    Capillary rise
    130 mm/10 min; 200 mm/30 min
  • Sheets
    460×570 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 262,00 €
    262,00 €
    1 * 50 ST
       262,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143271
    Grade
    BF 3
    Weight
    330 g/m²
    Thickness
    0,75 mm
    Capillary rise
    130 mm/10 min; 200 mm/30 min
  • Sheets
    580×600 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 348,00 €
    348,00 €
    1 * 50 ST
       348,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143273
    Grade
    BF 3
    Weight
    330 g/m²
    Thickness
    0,75 mm
    Capillary rise
    130 mm/10 min; 200 mm/30 min

Grade BF 4

Product Information

  • Dimensions
    Availability
    Price
  • Sheets
    150×150 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 45,90 €
    45,90 €
    1 * 25 ST
       45,90 €

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143287
    Grade
    BF 4
    Weight
    550 g/m²
    Thickness
    1,39 mm
    Capillary rise
    160 mm/10 min; 220 mm/30 min
  • Sheets
    580×600 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 236,00 €
    236,00 €
    1 * 25 ST
       236,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143296
    Grade
    BF 4
    Weight
    550 g/m²
    Thickness
    1,39 mm
    Capillary rise
    160 mm/10 min; 220 mm/30 min
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.