PES centrifugal concentrator, Vivaspin® 100
Supplier: Sartorius
Certificates
About this item
Vivaspin® 100 PES centrifugal concentrators are versatile ultrafiltration units for centrifugation-based sample concentration as well as pressure filtration applications.
- Easy to assemble and handle
- High flux PES membranes
- High target molecule recoveriesLargest capacity centrifugal device; for samples up to 90 ml
- Total process flexibility
- Constructed of PC body and filtrate vessel, PP concentrator cap
Popular applications for Vivaspin® centrifugal concentrators:
- Concentration | diafiltration of proteins, enzymes, nucleic acids, monoclonal antibodies, immunoglobulins, viruses and nanoparticles from samples up to 98 ml
- Concentration of cell culture supernatants
- Concentration of environmental samples
Specifications
- Catalog No:
- 516-0141
- 516-0146
- 516-0142
- 516-0143
- 516-0147
- 516-0144
- 516-0138
- 516-0148
- 516-0139
- 516-0140
- 516-0137
- 516-0145
- 516-0133
- 516-0136
- Filtration area:
- 23,5 cm²
- 23,5 cm²
- 23,5 cm²
- 23,5 cm²
- 23,5 cm²
- 23,5 cm²
- 23,5 cm²
- 23,5 cm²
- 23,5 cm²
- 23,5 cm²
- 23,5 cm²
- 23,5 cm²
- 23,5 cm²
- 23,5 cm²
- Max. RCF:
- 2000 ×g
- 2000 ×g
- 2000 ×g
- 2000 ×g
- 2000 ×g
- 2000 ×g
- 2000 ×g
- 2000 ×g
- 2000 ×g
- 2000 ×g
- 2000 ×g
- 2000 ×g
- 2000 ×g
- 2000 ×g
- Ø×H:
- 62×123/197 mm
- 62×123/197 mm
- 62×123/197 mm
- 62×123/197 mm
- 62×123/197 mm
- 62×123/197 mm
- 62×123/197 mm
- 62×123/197 mm
- 62×123/197 mm
- 62×123/197 mm
- 62×123/197 mm
- 62×123/197 mm
- 62×123/197 mm
- 62×123/197 mm
- Sample volume:
- 20 - 98 ml
- 20 - 100 ml
- 20 - 100 ml
- 20 - 98 ml
- 20 - 98 ml
- 20 - 100 ml
- 20 - 100 ml
- 20 - 98 ml
- 20 - 98 ml
- 20 - 100 ml
- 20 - 98 ml
- 20 - 98 ml
- 20 - 98 ml
- 20 - 100 ml
- Dead stop volume:
- 350 µl
- 350 µl
- 350 µl
- 350 µl
- 350 µl
- 350 µl
- 350 µl
- 350 µl
- 350 µl
- 350 µl
- 350 µl
- 350 µl
- 350 µl
- 350 µl
- Hold-up volume:
- < 250 µl
- < 250 µl
- < 250 µl
- < 250 µl
- < 250 µl
- < 250 µl
- < 250 µl
- < 250 µl
- < 250 µl
- < 250 µl
- < 250 µl
- < 250 µl
- < 250 µl
- < 250 µl