Disposable earplugs, Laser Lite®
Supplier: HONEYWELL SAFETY
Jump to:
About this item
Laser Lite® earplugs offer optimal comfort for long-term wear.
- Bright colours for high visibility
- Self-adjusting foam to fit virtually every user
- T-shape for easy insertion and wear
SNR: 35 dB
EN 352-2
Certifications: EN 352-2
Caution: * One size fits all.
3 Options Available Below
Product Details & Documents
Laser Lite® earplugs offer optimal comfort for long-term wear.
- Bright colours for high visibility
- Self-adjusting foam to fit virtually every user
- T-shape for easy insertion and wear
SNR: 35 dB
EN 352-2
Certifications: EN 352-2
Caution: * One size fits all.
Product Family Options
Product Information
- ColourSizeAvailabilityPrice
- 111-0137Earplugs, without cord, packed in pairs
- 111-0138Earplugs, with cord, packed in pairs
- 111-0395Earplugs without cord, bulk packaging, refill for dispenser LS-400Earplugs without cord, bulk packaging, refill for dispenser LS-400Yellow/redU*Log in for availability1 * 200 PAAR48,20 €