VWR®, Soap Dispensers, Touch-Free
Catalog # 129-0486
Supplier: VWR Collection
Specifications
- Colour:White
- Description:Wall-mounting soap/gel dispenser, Touch-Free
- Pk:1
Specifications
About this item
The Touch-Free soap dispenser reduces cross-contamination and waste in cleanrooms and supporting areas.
- No-waste unit, wall-mounting
- Electronic eye to activate a measured dose of soap
- Compact gravity-fed unit without tube but with a pump action
Suitable for use with Micronova 500 ml pump bottles.
Certifications: OSHA, CDC, ADA
Delivery: Supplied with four size 'D' batteries.