ContactUsLinkComponent
Glove box holder
Glove box holder
Catalog # 112-1529
undefined
Glove box holder
Catalog # 112-1529
Specifications
  • Colour:
    White
  • Description:
    Glove box holder
  • W×D×H:
    245×84×145 mm
  • Pk:
    1
Specifications
About this item
White boxes holder to simply insert from the side the gloves which can be taken out from the top.
  • Acrylic solid and modern design
  • Hold glove boxes with 100 gloves
  • For use on a table or wall-mounted