VWR®, Dishes, Evaporating, Shallow
Porcelain, glazed.
- Very good chemical resistance
- Heat resistant up to +1000 °C
- Low thermal expansion
The economical alternative for a wide range of laboratory applications. With spout and flat bottom. Glazed white except for the outside bottom surface.
DIN 12903
: DIN 12903