Orders LinkContactUsLinkComponent
Dishes, evaporating
Dishes, evaporating
Dishes, evaporating
Certificates
Dishes, evaporating
Dishes, evaporating
6 Options Available Below
Product Details & Documents
Porcelain, glazed.
  • Very good chemical resistance
  • Heat resistant up to +1000 °C
  • Low thermal expansion
For a wide range of laboratory applications. With spout and flat bottom. White glaze inside and outside.
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Availability
    Price
  • 30 ml
    24 mm
    58 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,60 €
    16,60 €
    1 * 1 ST
       16,60 €
  • 60 ml
    30 mm
    75 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 27,90 €
    27,90 €
    1 * 1 ST
       27,90 €
  • 100 ml
    36 mm
    84 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,10 €
    34,10 €
    1 * 1 ST
       34,10 €
  • 150 ml
    40 mm
    97 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,80 €
    42,80 €
    1 * 1 ST
       42,80 €
  • 250 ml
    50 mm
    110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 53,00 €
    53,00 €
    1 * 1 ST
       53,00 €
  • 400 ml
    60 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 74,20 €
    74,20 €
    1 * 1 ST
       74,20 €