Orders LinkContactUsLinkComponent
Dilatometers, Quickfit®
Dilatometers, Quickfit®
Supplier:   SciLabware
Dilatometers, Quickfit®
Supplier:  SciLabware
Certificates
Dilatometers, Quickfit®
Dilatometers, Quickfit®
2 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass.
  • Dilatometer body is graduated with an arbitrary scale
  • Interchangable stopper has open top to allow weighting with steel balls etc.
For measuring the expansion of edible oils and fats over the temperature range within which melting takes place.
Ordering information: The body and stopper should be ordered separately.
Product Family Options
Product Information
  • Availability
    Price
  • BIBBWE12/1
    Dilatometer body, with socket size NS 14/23, without stopper
    Dilatometer body, with socket size NS 14/23, without stopper
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 1 060,00 €
    1 060,00 €
    1 * 5 ST
       1 060,00 €
  • BIBBSB14DL
    Dilatometer stopper with cone size NS 14/23
    Dilatometer stopper with cone size NS 14/23
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 176,00 €
    176,00 €
    1 * 5 ST
       176,00 €