ContactUsLinkComponent
Desiccator cabinets
Catalog # 467-0004
Supplier:  SciLabware
undefined
Desiccator cabinets
Catalog # 467-0004
Supplier:  SciLabware
Specifications
  • W×D×H ext.:
    200×225×168 mm
  • Description:
    Polystyrene desiccator cabinet
  • Pk:
    1
  • With:
    door opening 188×110 (mm)
Specifications
About this item
Clear, high impact polystyrene cabinets have an airtight door with foam rubber gasket.
  • Cabinets can be stacked to save space
Delivery: Supplied with sealed jar of silica gel.