Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Crucibles
Crucibles, low form
Crucibles, low form
:
TECHNICAL QUARTZ SOLUTIONS
Crucibles, low form
:
TECHNICAL QUARTZ SOLUTIONS
These crucibles are made from fused quartz, translucent, glazed.
Max. temperature: 1050 °C
:
Crucible and lid have to be ordered separately.
Lids