Crucibles, porcelain
Supplier: MORGAN ADVANCED MATERIALS HALD
Certificates
About this item
Porcelain, glazed inside and out, except base.
- Withstand maximum temperature of 1000 °C
Lids are available separately.
Porcelain, glazed inside and out, except base.
Lids are available separately.