Crucibles, metal
These crucibles are made from iron.
- Recommended maximum temperature 1100 °C
- Can be used up to 1500 °C despite a risk of shortening the life time
Area of use: Only on open burner flame.
: Lids are available separately.
These crucibles are made from iron.
Area of use: Only on open burner flame.