ContactUsLinkComponent
One-piece hex-head fittings
One-piece hex-head fittings
Catalog # 554-1154
Supplier:  VICI JOUR RESEARCH
One-piece hex-head fittings
Catalog # 554-1154
Supplier:  VICI JOUR RESEARCH
Specifications
  • Description:
    One-piece 10-32 hex-head fitting, long
  • Pk:
    6
Specifications
About this item
Peek one-piece hex-head fittings, with their integrated ferrule, are easy to use with the valve-tool and the colour-it colour-coded finger-tight adapters. PEEK one-piece hex-head fittings are available in short and long versions.
  • Ideal for use with PEEK tubing
  • 10-32 thread will fit virtually all standard high-pressure female connection
Withstand pressures up to 350 bar (5000 psi)
Withstand pressures up to 350 bar (5000 psi)