HPLC columns, ZirChrom®
Supplier: Avantor
Certificates
About this item
Zirconia bonded phases offer unique selectivity relative to conventional silica based columns, combined with extreme chemical and thermal stability. Unlike polymeric phases, zirconia will not shrink or swell as a function of eluent organic content or ionic strength.
- Porous zirconia based reverse phase materials
- High pH and temperature stability
- Fast solvent equilibration
Frequently Bought Together