Size exclusion chromatography columns, GPC KF-800 series
: Resonac
A complete range of general purpose gel permeation columns (GPC) packed with tetrahydrofuran (THF). Columns packed with other solvents are also available. Please enquire for further details.
- Column types KF-801 to 802.5 for compounds with a molecular weight of less than 2000 g.mol⁻¹, such as liposoluble vitamins (KF-801), oligomers (KF-802-803) and polymer additives (KF-801-802)
- Column types KF-802.5 to 807L for compounds with a molecular weight of 2000 g.mol⁻¹ or more, such as THF-soluble synthetic polymers (KF803-807L) and oligomers (KF802-803)
- L represents a 'linear column' which exhibits linearity in the calibration curve over a wide range of molecular weights
* Estimated value
- Catalog No:
- 554-0866
- 554-0877
- 554-0875
- 554-1765
- 554-0869
- 554-0871
- 554-0868
- 554-0865
- 554-0867
- 554-0874
- 554-0864
- 554-2429
- 554-0870
- 554-0872
- Description:
- GPC KF-802.5
- GPC KF-807L
- GPC KF-806M
- GPC KF-G 4A
- GPC KF-804
- GPC KF-805
- GPC KF-803L
- GPC KF-802
- GPC KF-803
- GPC KF-806L
- GPC KF-801
- GPC KF-800D
- GPC KF-804L
- GPC KF-805L
- Max. pressure:
- 3,5 MPa (35 bar)
- 3,5 MPa (35 bar)
- 3,5 MPa (35 bar)
- 3,5 MPa (35 bar)
- 3,5 MPa (35 bar)
- 3,5 MPa (35 bar)
- 3,5 MPa (35 bar)
- 3,5 MPa (35 bar)
- 3,5 MPa (35 bar)
- 3,5 MPa (35 bar)
- 3,5 MPa (35 bar)
- 3,5 MPa (35 bar)
- Temperature limit:
- ≤60 °C
- ≤60 °C
- ≤60 °C
- ≤60 °C
- ≤60 °C
- ≤60 °C
- ≤60 °C
- ≤60 °C
- ≤60 °C
- ≤60 °C
- ≤60 °C
- ≤60 °C
- ≤60 °C
- ≤60 °C
- USP code:
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21