Sleeve protectors, Tyvek® 500, model PS32LA
Catalog # 113-0494
Supplier: DuPont
Specifications
- Garment application:Pharmaceutical,Laboratory
- Colour:White
- Length:500 mm
- Pk:200
Specifications
About this item
Specially designed for use with Tyvek® apparel, these sleeve protectors can help offer enhanced protection for body parts that are more exposed to hazardous substances. Tyvek® sleeves are made from flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort.
- Ultra-low linting fabric
- Adjustable arm opening
- Stitched internal seams
- Upper-arm in blue-coloured thread for identification purposes
- Anti-static treatment (EN 1149-5) - on both sides
The fabric is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 µm in size). Typical applications include: pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up.
Weight: 41 g/m²
Certifications: (EU) 2016/425; Cat III, Type PB [6-B]; EN 14126
Packaging: 200 per box, bulk packed.