- Garment application:Chemical protection,Oils, fats and fuels,Industrial
- Colour:Grey
- Pk:50
Specially designed for use with Tychem® apparel, these boot covers can help offer enhanced protection for body parts that are more exposed to hazardous substances.
- Knee-length boot covers with slip-retardant sole and fixation ties
- Sole is partially stitched: splash-proof, not fully liquid tight
- Stitched and over-taped seams with barrier tape for protection and strength
- Anti-static treatment (EN 1149-5) inside only
- Conform to EN 14126 (barrier to infective agents)
Tychem® F boot covers are made from a lightweight and durable fabric consisting of a proprietary barrier film laminated to a heavy duty Tyvek® substrate. They offer excellent chemical permeation protection to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents. Ideal for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications.
Weight: 120 g/m²