Cleanroom shoe covers, PP
113-0408
- Shoe size:U*
- Garment application:Cleanroom
- Colour:Blue
- Length:445 mm
- Pk:300
These anti-skid shoe covers made from spunbonded PP may be used to protect against dirt, grime, and certain dry particulates in non hazardous environments.
- Appropriate for industrial, pharmaceutical, food processing, construction, and certain environmental cleanup applications
- Elastic top and bottom
*One size fits all