Shoe covers, Tyvek® 500, models POS0 / POSA
Supplier: DuPont
Certificates
About this item
Specially designed for use with Tyvek® apparel, these Tyvek® 500 shoe covers can help offer enhanced protection for body parts that are more exposed to hazardous substances.
- Ultra-low linting fabric
- Elasticated ankle
- Stitched internal seams
- Anti-static treatment (EN 1149-5) on both sides (excludes slip-retardant sole)
- Two styles available: with or without slip-retardant sole
Tyvek® is composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort. The fabric is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 µm in size). Typical applications include pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up, amongst many others.
Weight: 41 g/m²
One size fits all.
Certifications: (EU) 2016/425; Cat III, Type PB [6-B]; EN 14126.
Packaging: POS0 and POSA models come 200 and 400 per box in bulk packaging respectively.
Specifications
- Catalog No:
- 113-0493
- 113-0492
- 113-0491
- Garment application:
- Laboratory
- Laboratory
- Laboratory