Aprons, Tyvek® 500, model PA30L0
Catalog # 113-0479
Supplier: DuPont
Specifications
- Anti-static:+
- Garment application:Chemical protection,General Purpose,Pharmaceutical,Industrial
- Colour:White
- Pk:100
Specifications
About this item
The apron is made of Tyvek®, a fabric composed of flash spun high density PE. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 micron in size). Specially designed for use with Tyvek® apparel, this apron offers enhanced protection for body parts that are more exposed to hazardous substances.
- Shin-length apron with neck and waist ties
- Ultra-low-linting fabric
- Antistatic treatment (EN 1149-5) - on both sides
- No silicone added
Typical applications are pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up.
Length: 108 cm
Certifications: Type PB [6-B]; EN 14126; Cat. III; (EU) 2016/425
Packaging: 100 per box, bulk packed.