Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Closures
Bottle Closures
Screw caps
Screw caps
Screw caps
Supplier:
Kautex
Certificates
Expand image
Screw caps
Supplier:
Kautex
About this item
LDPE, natural.
Good chemical resistance
Good thermal resistance
Leakproof
Related Information
Documentation
Videos
Kautex Wide Neck Bottle Series 303 LDPE
Documentation