Orders LinkContactUsLinkComponent
Replacement screw caps
Replacement screw caps
Replacement screw caps
Certificates
Replacement screw caps
Replacement screw caps
2 Options Available Below
Product Details & Documents
PP, black.
  • Very good chemical and thermal resistance
  • Suitable for bottles 215-1821 to 215-1836 and culture bottle 215-1861 to 215-1866
  • LUP lining for better sealing
Product Family Options
Product Information
  • Thread
    For bottles
    Availability
    Price
  • GL 45
    1000 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 88,80 €
    88,80 €
    1 * 100 ST
       88,80 €
  • GL 68
    1000 ml
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 19,40 €
    19,40 €
    1 * 25 ST
       19,40 €